3-Methylcyclopentan-1,2-dion ≥98%

Lieferant: Thermo Fisher Scientific

A10255.09 A10255.18 A10255.22
A10255.09EA 25.8 EUR
A10255.09 A10255.18 A10255.22
3-Methylcyclopentan-1,2-dion ≥98%
3-Methylcyclopentan-1,2-dion

Purity: 98+% CAS No.: 765-70-8 Molecular Formula: C6H8O2 Formula Weight: 112.13 Melting Point: 103-107° Beilstein Reference: 2076520

Formel: C₆H₈O₂
Siedepunkt: 210 °C (1013 hPa)
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001417
CAS-Nummer: 765-70-8
EINECS: 212-154-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR