5-Chlor-2-pentanon ≥94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 94%
CAS No.: 5891-21-4
Molecular Formula: C5H9ClO
Formula Weight: 120.58
Storage Temperature: 2°C to 8°C
Flash Point: 35°(95°F)
Refractive Index: 1.4380
Boiling Point: 63-64°/12mm
Beilstein Reference: 1098490
UN No.: 1224
MDL No.: MFCD00001008
Formel:
C₅H₉ClO Siedepunkt: 63…64 °C (12 mmHg) Schmelzpunkt: –48 °C Dichte: 1,053 Flammpunkt: 35 °C (95 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001008 CAS-Nummer: 5891-21-4 EINECS: 227-565-8 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...