cis-3-Hexen-1-ol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10313.09
A10313.18
A10313.30
A10313.09EA
44.8
EUR
A10313.09
A10313.18
A10313.30
cis-3-Hexen-1-ol ≥98%
cis-3-Hexen-1-ol
Purity: 98%
CAS No.: 928-96-1
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 44°(111°F)
Refractive Index: 1.4400
Odour: Pleasant
Boiling Point: 156-157°
Beilstein Reference: 1719712
Merck Reference: 14,4700
UN No.: 1987
MDL No.: MFCD00063217
Formel:
C₆H₁₂O Siedepunkt: 156…157 °C Schmelzpunkt: –60 °C Dichte: 0,848 Flammpunkt: 44 °C (111 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063217 CAS-Nummer: 928-96-1 EINECS: 213-192-8 UN: 1987 ADR: 3,III Merck Index: 14,4700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...