Bariumacetat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Essigsäure Bariumsalz, Bariumdi(acetat)
A10316.30
A10316.30EA
29.3
EUR
A10316.30
Bariumacetat ≥99%
Bariumacetat
Purity: 99%
CAS No.: 543-80-6
Molecular Formula: Ba(OOCCH3)2
Formula Weight: 255.43
Odour: Odourless
Merck Reference: 14,965
MDL No.: MFCD00012447
Formel:
(H₃CCOO)₂Ba Molecular Weight: 255.42 g/mol Schmelzpunkt: ∼450 °C Dichte: 2.47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012447 CAS-Nummer: 543-80-6 EINECS: 208-849-0 UN: 1564 ADR: 6.1 III Merck Index: 13,00968 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...