n-Hexadecan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cetan
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.
Purity: 99%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colourless
Melting Point: 18°
Boiling Point: 287°
Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water.
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142
Formel:
H₃C(CH₂)₁₄CH₃ Molecular Weight: 226.45 g/mol Siedepunkt: 287 °C Schmelzpunkt: 18 Dichte: 0.773 Flammpunkt: 135 °C (275 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008998 CAS-Nummer: 544-76-3 EINECS: 208-878-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...