4,4'-Isopropylidendiphenol 97
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propan
A10324.0B
A10324.0I
A10324.30
A10324.0BEA
48.6
EUR
A10324.0B
A10324.0I
A10324.30
4,4'-Isopropylidendiphenol 97
4,4'-Isopropylidendiphenol
Purity: 97+%
CAS No.: 80-05-7
Molecular Formula: C15H16O2
Formula Weight: 228.29
Flash Point: 227°(440°F)
Melting Point: 153-158°
Boiling Point: 200°/4mm
Beilstein Reference: 1107700
Merck Reference: 14,1297
MDL No.: MFCD00002366
Formel:
C₁₅H₁₆O₂ Molecular Weight: 228.29 g/mol Siedepunkt: 200 °C (4 mmHg) Schmelzpunkt: 153…158 °C Dichte: 1.205 g/cm³ (20 °C) Flammpunkt: 227 °C (440 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002366 CAS-Nummer: 80-05-7 EINECS: 201-245-8 Merck Index: 13,01296 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...