N,N-Dimethyl-1,4-phenylendiammoniumdichlorid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10343.36 A10343.22 A10343.14
A10343.36EA 967 EUR
A10343.36 A10343.22 A10343.14
N,N-Dimethyl-1,4-phenylendiammoniumdichlorid 98%
N,N-Dimethyl-1,4-phenylendiammoniumdichlorid

Purity: 98% CAS No.: 536-46-9 Molecular Formula: C8H12N2.2HCl Formula Weight: 209.12 Melting Point: 212° dec. Solubility: Soluble in water MDL No.: MFCD00012991

Formel: (CH₃)₂NC₆H₄NH₂·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012991
CAS-Nummer: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR