Morpholin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 110-91-8
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 1.007
Refractive Index: 1.4540
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Amine-like
Melting Point: -5°
Boiling Point: 129°
Beilstein Reference: 102549
Merck Reference: 14,6277
UN No.: 2054
MDL No.: MFCD00005972
Formel:
C₄H₉NO Siedepunkt: 129 °C Schmelzpunkt: –5 °C Dichte: 1,007 Flammpunkt: 35 °C (95 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005972 CAS-Nummer: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8,I Merck Index: 14,6277 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...