L(+)-Cystein Hydrochlorid Monohydrat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-(+)-Cystein Hydrochlorid Monohydrat, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid Monohydrat, H-Cys-OH.HCl.H2O

A10389.14 A10389.22 A10389.36
A10389.14EA 32 EUR
A10389.14 A10389.22 A10389.36
L(+)-Cystein Hydrochlorid Monohydrat ≥99%
L(+)-Cystein Hydrochlorid Monohydrat

Purity: 99% CAS No.: 7048-04-6 Molecular Formula: C3H7NO2S.HCl.H2O Formula Weight: 175.64 (157.62anhy) pH: 1-2 Melting Point: ca 175° dec. Beilstein Reference: 5158059 Merck Reference: 14,2781 Optical Rotation: +5.5° (c=5 in 5N HCl) MDL No.: MFCD00065606

Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
Schmelzpunkt: ∼ 175 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00065606
CAS-Nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 14,2781
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR