2,2,3,3,4,4,4-Heptafluor-1-butanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10414.09
A10414.14
A10414.18
A10414.09EA
66.5
EUR
A10414.09
A10414.14
A10414.18
2,2,3,3,4,4,4-Heptafluor-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluor-1-butanol
Purity: 98%
CAS No.: 375-01-9
Molecular Formula: C4H3F7O
Formula Weight: 200.06
Flash Point: 25°(77°F)
Refractive Index: 1.300
Boiling Point: 95-97°
Beilstein Reference: 1761907
UN No.: 1987
MDL No.: MFCD00004674
Formel:
C₄H₃F₇O Siedepunkt: 95…97 °C Dichte: 1,596 Flammpunkt: 25 °C (77 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004674 CAS-Nummer: 375-01-9 EINECS: 206-782-1 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...