Cholesteryl chloride 98%
Lieferant: Thermo Fisher Scientific
A10433.14
A10433.22
A10433.14EA
60
EUR
A10433.14
A10433.22
Cholesteryl chloride 98%
Cholesteryl chloride
Purity: 98%
CAS No.: 910-31-6
Molecular Formula: C27H45Cl
Formula Weight: 405.11
Melting Point: 94-96°
Beilstein Reference: 2703655
Optical Rotation: -26° (c=1 in benzene)
MDL No.: MFCD00003632
Formel:
C₂₇H₄₅Cl Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003632 CAS-Nummer: 910-31-6 EINECS: 213-004-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...