L(+)-Cystein ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98+%
CAS No.: 52-90-4
Molecular Formula: C3H7NO2S
Formula Weight: 121.16
Physical Form: Crystalline powder
Sensitivity: Air Sensitive
Appearance: White
Odour: Odourless
Melting Point: ca 220° dec.
Beilstein Reference: 1721408
Merck Reference: 14,2781
Optical Rotation: +7.9° (c=5 in 5N HCl)
MDL No.: MFCD00064306
Formel:
HSCH₂CH(NH₂)CO₂H Siedepunkt: 254 °C (1013 hPa) Dichte: 1,523 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064306 CAS-Nummer: 52-90-4 EINECS: 200-158-2 Merck Index: 14,2781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...