4'-Hydroxy-3'-methoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10439.18 A10439.30 A10439.36
A10439.18EA 48.1 EUR
A10439.18 A10439.30 A10439.36
4'-Hydroxy-3'-methoxyacetophenon ≥98%
4'-Hydroxy-3'-methoxyacetophenon

Purity: 98% CAS No.: 498-02-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 112-116° Boiling Point: 295-300° Merck Reference: 14,741 MDL No.: MFCD00008747

Formel: C₉H₁₀O₃
Siedepunkt: 295…300 °C
Schmelzpunkt: 112…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008747
CAS-Nummer: 498-02-2
EINECS: 207-854-5
Merck Index: 14,741

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR