2-Amino-6-methylpyridin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
6-Methylpyridin-2-amin, 2-Amino-6-picolin, 6-Methyl-2-pyridylamin
A10444.22
A10444.36
A10444.22EA
40.6
EUR
A10444.22
A10444.36
2-Amino-6-methylpyridin 99%
2-Amino-6-methylpyridin
Purity: 99%
CAS No.: 1824-81-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 103°(217°F)
Odour: Unpleasant
Boiling Point: 208-209°
Beilstein Reference: 107048
UN No.: 2811
MDL No.: MFCD00006331
Formel:
C₆H₈N₂ Molecular Weight: 108.14 g/mol Siedepunkt: 208…209 °C Schmelzpunkt: 41…45 °C Flammpunkt: 103 °C (217 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006331 CAS-Nummer: 1824-81-3 EINECS: 217-360-1 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...