2,6-Lutidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,6-Dimethylpyridine
A10478.0F
A10478.AC
A10478.AE
A10478.AP
A10478.0FEA
340
EUR
A10478.0F
A10478.AC
A10478.AE
A10478.AP
2,6-Lutidin ≥98%
2,6-Lutidin
Purity: 98+%
CAS No.: 108-48-5
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 33°(91°F)
Refractive Index: 1.4970
Odour: Unpleasant
Melting Point: -7°
Boiling Point: 143-145°
Beilstein Reference: 105690
Merck Reference: 14,5616
UN No.: 1993
MDL No.: MFCD00006345
Formel:
C₇H₉N Siedepunkt: 143…145 °C Schmelzpunkt: –7 °C Dichte: 0,923 Flammpunkt: 33 °C (91 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006345 CAS-Nummer: 108-48-5 EINECS: 203-587-3 UN: 1993 ADR: 3,III Merck Index: 14,5616 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...