1,2-Naphthochinon-4-sulfonsäure Natriumsalz ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 521-24-4
Molecular Formula: C10H5NaO5S
Formula Weight: 260.20
Melting Point: ca 289° dec.
Solubility: Soluble in water. Moderately soluble in acetone. Slightly soluble in alcohol
Beilstein Reference: 4169788
Merck Reference: 14,8646
MDL No.: MFCD00001700
Formel:
C₁₀H₅NaO₅S Schmelzpunkt: ∼ 289 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001700 CAS-Nummer: 521-24-4 EINECS: 208-308-9 Merck Index: 14,8646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...