2-Chlor-6-fluorbenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10513.14
A10513.22
A10513.14EA
29
EUR
A10513.14
A10513.22
2-Chlor-6-fluorbenzaldehyd ≥97%
2-Chloro-6-fluorobenzaldehyde
Purity: 97%
CAS No.: 387-45-1
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 101°(213°F)
Melting Point: 35-39°
Boiling Point: 91-93°/10mm
Beilstein Reference: 2245530
MDL No.: MFCD00003306
Formel:
C₇H₄ClFO Siedepunkt: 91…93 °C (10 mmHg) Schmelzpunkt: 35…39 °C Flammpunkt: 101 °C (213 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003306 CAS-Nummer: 387-45-1 EINECS: 206-860-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...