1,4-Dibrombenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10517.18 A10517.0E A10517.22 A10517.30
A10517.18EA 22.8 EUR
A10517.18 A10517.0E A10517.22 A10517.30
1,4-Dibrombenzol ≥98%
1,4-Dibrombenzol

Purity: 98% CAS No.: 106-37-6 Molecular Formula: C6H4Br2 Formula Weight: 235.91 Flash Point: 100°(212°F) Physical Form: Crystalline powder Density: 1.841 Refractive Index: 1.5742 Appearance: White Odour: Characteristic Melting Point: 86-90° Boiling Point: 219-220° Beilstein Reference: 1904543 Merck Reference: 14,3019 UN No.: 3077 MDL No.: MFCD00000089

Formel: C₆H₄Br₂
Siedepunkt: 219…220 °C
Schmelzpunkt: 86…90 °C
Dichte: 1,841
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000089
CAS-Nummer: 106-37-6
EINECS: 203-390-2
UN: 3077
ADR: 9,III
Merck Index: 14,3019

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR