Valerophenon ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Phenyl-1-pentanon

A10525.14 A10525.22 A10525.36
A10525.14EA 43.5 EUR
A10525.14 A10525.22 A10525.36
Valerophenon ≥99%
Valerophenon

Purity: 99% CAS No.: 1009-14-9 Molecular Formula: C11H14O Formula Weight: 162.23 Flash Point: 102°(215°F) Refractive Index: 1.5143 Melting Point: -9° Boiling Point: 244-245° Beilstein Reference: 1907717 MDL No.: MFCD00009480

Formel: C₆H₅C(O)(CH₂)₃CH₃
Molecular Weight: 162.23 g/mol
Siedepunkt: 244…245 °C
Schmelzpunkt: –9 °C
Dichte: 0.978
Flammpunkt: 102 °C (215 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009480
CAS-Nummer: 1009-14-9
EINECS: 213-767-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2584f16b192b481fbaf1d8ab1e77be6109fb67a6&vid=3bf6c22512cdad990d29bc1ffe3db3b535a55970&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR