Methyl-3-methyl-2-furoat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl 3-methylfuran-2-carboxylate
A10541.14
A10541.06
A10541.09
A10541.14EA
250
EUR
A10541.14
A10541.06
A10541.09
Methyl-3-methyl-2-furoat ≥98%
Methyl-3-methyl-2-furoat
Purity: 98%
CAS No.: 6141-57-7
Molecular Formula: C7H8O3
Formula Weight: 140.14
Flash Point: 82°(180°F)
Melting Point: 33-37°
Boiling Point: 60°/1mm
Beilstein Reference: 116835
MDL No.: MFCD00014110
Formel:
C₇H₈O₃ Siedepunkt: 60 °C (1 mmHg) Schmelzpunkt: 33…37 °C Flammpunkt: 82 °C (180 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014110 CAS-Nummer: 6141-57-7 EINECS: 228-131-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...