Methyl-3-methyl-2-furoat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 3-methylfuran-2-carboxylate

A10541.14 A10541.06 A10541.09
A10541.14EA 250 EUR
A10541.14 A10541.06 A10541.09
Methyl-3-methyl-2-furoat ≥98%
Methyl-3-methyl-2-furoat

Purity: 98% CAS No.: 6141-57-7 Molecular Formula: C7H8O3 Formula Weight: 140.14 Flash Point: 82°(180°F) Melting Point: 33-37° Boiling Point: 60°/1mm Beilstein Reference: 116835 MDL No.: MFCD00014110

Formel: C₇H₈O₃
Siedepunkt: 60 °C (1 mmHg)
Schmelzpunkt: 33…37 °C
Flammpunkt: 82 °C (180 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014110
CAS-Nummer: 6141-57-7
EINECS: 228-131-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR