Methyl-4-iod-2-methoxybenzoat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Iod-2-methoxybenzoesäuremethylester

A10549.06 A10549.14
A10549.06EA 47.1 EUR
A10549.06 A10549.14
Methyl-4-iod-2-methoxybenzoat ≥98%
Methyl-4-iod-2-methoxybenzoat

Purity: 98% CAS No.: 35387-93-0 Molecular Formula: C9H9IO3 Formula Weight: 292.07 Melting Point: 93-95° Beilstein Reference: 2692711

Formel: C₉H₉IO₃
Molecular Weight: 292.07 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052925
CAS-Nummer: 35387-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR