N,N-Diethylanilin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A10560.0D A10560.0F A10560.AP
A10560.0DEA 352 EUR
A10560.0D A10560.0F A10560.AP
N,N-Diethylanilin ≥99%
N,N-Diethylanilin

Purity: 99% CAS No.: 91-66-7 Molecular Formula: C10H15N Formula Weight: 149.24 Flash Point: 85°(185°F) Refractive Index: 1.5420 Melting Point: -38° Boiling Point: 215-216° Beilstein Reference: 742483 Merck Reference: 14,3114 UN No.: 2432 MDL No.: MFCD00009042

Formel: C₁₀H₁₅N
Siedepunkt: 215…216 °C
Schmelzpunkt: –38 °C
Dichte: 0,936
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009042
CAS-Nummer: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Merck Index: 14,3114

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR