Dimethyl-2-aminoterephthalat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

A10567.0B A10567.18 A10567.30
A10567.0BEA 614 EUR
A10567.0B A10567.18 A10567.30
Dimethyl-2-aminoterephthalat 99%
Dimethyl-2-aminoterephthalat

Purity: 99% CAS No.: 5372-81-6 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 130-134° Beilstein Reference: 2808505

Formel: C₁₀H₁₁NO₄
Schmelzpunkt: 130…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008427
CAS-Nummer: 5372-81-6
EINECS: 226-364-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR