Nitrobenzol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 98-95-3
Molecular Formula: C6H5NO2
Formula Weight: 123.11
Flash Point: 87°(188°F)
Refractive Index: 1.5520
Melting Point: 5-6°
Boiling Point: 210-212°
Beilstein Reference: 507540
Merck Reference: 14,6588
UN No.: 1662
MDL No.: MFCD00007043
Formel:
C₆H₅NO₂ Siedepunkt: 210…212 °C Schmelzpunkt: 5…6 °C Dichte: 1,196 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007043 CAS-Nummer: 98-95-3 EINECS: 202-716-0 UN: 1662 ADR: 6.1,II Merck Index: 14,6588 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...