Tetrabutylammoniumfluorid 1 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10588.AE
A10588.AC
A10588.AP
A10588.AEEA
114
EUR
A10588.AE
A10588.AC
A10588.AP
Tetrabutylammoniumfluorid 1 M in THF
Tetrabutylammoniumfluorid
CAS No.: 429-41-4
Molecular Formula: C16H36FN
Formula Weight: 261.47
Flash Point: -17°(1°F)
Beilstein Reference: 3570522
UN No.: 2924
MDL No.: MFCD00011747
Formel:
C₁₆H₃₆FN Dichte: 0,903 Flammpunkt: –17 °C (1 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011747 CAS-Nummer: 429-41-4 EINECS: 207-057-2 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...