Dimethyl-5-nitroisophthalat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Nitro-1,3-benzoldicarbonsäuredimethylester, 5-Nitroisophthalsäuredimethylester

A10610.0B A10610.30
A10610.0BEA 213 EUR
A10610.0B A10610.30
Dimethyl-5-nitroisophthalat ≥98%
Dimethyl-5-nitroisophthalat

Purity: 98+% CAS No.: 13290-96-5 Molecular Formula: C10H9NO6 Formula Weight: 239.19 Melting Point: 121-123° Beilstein Reference: 2140916

Formel: C₁₀H₉NO₆
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008429
CAS-Nummer: 13290-96-5
EINECS: 236-307-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR