1-Brom-2-fluorbenzol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 1072-85-1
Molecular Formula: C6H4BrF
Formula Weight: 175.01
Flash Point: 43°(109°F)
Refractive Index: 1.5340
Odour: Aromatic
Melting Point: -8°
Boiling Point: 155-157°
Beilstein Reference: 1071819
UN No.: 1993
MDL No.: MFCD00000282
Formel:
C₆H₄BrF Siedepunkt: 155…157 °C Schmelzpunkt: –8 °C Dichte: 1,601 Flammpunkt: 43 °C (109 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000282 CAS-Nummer: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...