3-Fluor-o-xylol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10637.06
A10637.14
A10637.22
A10637.06EA
30.5
EUR
A10637.06
A10637.14
A10637.22
3-Fluor-o-xylol ≥99%
3-Fluor-o-xylol
Purity: 99%
CAS No.: 443-82-3
Molecular Formula: C8H9F
Formula Weight: 124.16
Flash Point: 36°(96°F)
Refractive Index: 1.4860
Odour: Aromatic
Boiling Point: 147-148°
Beilstein Reference: 2040955
UN No.: 1993
MDL No.: MFCD00000323
Formel:
C₈H₉F Siedepunkt: 147…148 °C Dichte: 0,992 Flammpunkt: 36 °C (96 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000323 CAS-Nummer: 443-82-3 EINECS: 207-140-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...