Methyl-3-(methylthio)propionat ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Methyl3-(methylthio)propionate
A10649.14
A10649.22
A10649.36
A10649.14EA
29
EUR
A10649.14
A10649.22
A10649.36
Methyl-3-(methylthio)propionat ≥98%
Methyl-3-(methylthio)propionat
Purity: 98%
CAS No.: 13532-18-8
Molecular Formula: C5H10O2S
Formula Weight: 134.20
Flash Point: 72°(161°F)
Refractive Index: 1.4650
Boiling Point: 72-73°/11mm
Beilstein Reference: 1745077
MDL No.: MFCD00008565
Formel:
C₅H₁₀O₂S Siedepunkt: 72…73 °C (11 mmHg) Dichte: 1,074 Flammpunkt: 72 °C (161 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008565 CAS-Nummer: 13532-18-8 EINECS: 236-883-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...