Isonicotinaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 872-85-5
Molecular Formula: C6H5NO
Formula Weight: 107.11
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.5440
Melting Point: -4°
Boiling Point: 198°
Beilstein Reference: 105342
MDL No.: MFCD00006425
Formel:
C₆H₅NO Siedepunkt: 198 °C Schmelzpunkt: –4 °C Dichte: 1,132 Flammpunkt: 75 °C (167 °F) |
MDL:
MFCD00006425 CAS-Nummer: 872-85-5 EINECS: 212-832-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...