2-Amino-3-hydroxypyridin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10665.18 A10665.09 A10665.22 A10665.30
A10665.18EA 70 EUR
A10665.18 A10665.09 A10665.22 A10665.30
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin

Purity: 98% CAS No.: 16867-03-1 Molecular Formula: C5H6N2O Formula Weight: 110.12 Odour: Odourless Melting Point: 166-174° Beilstein Reference: 109868 UN No.: 2811 MDL No.: MFCD00006317

Formel: C₅H₆N₂O
Siedepunkt: 250 °C (1013 hPa)
Schmelzpunkt: 166…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006317
CAS-Nummer: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR