4-(Trifluormethoxy)benzaldehyd 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10671.14
A10671.06
A10671.14EA
147
EUR
A10671.14
A10671.06
4-(Trifluormethoxy)benzaldehyd 96%
4-(Trifluormethoxy)benzaldehyd
Purity: 96%
CAS No.: 659-28-9
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Flash Point: 71°(159°F)
Refractive Index: 1.4580
Boiling Point: 93°/27mm
Beilstein Reference: 1949135
MDL No.: MFCD00041530
Formel:
C₈H₅F₃O₂ Siedepunkt: 93 °C (27 mmHg) Dichte: 1,331 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041530 CAS-Nummer: 659-28-9 EINECS: 211-531-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...