Phenylglyoxal monohydrate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone

A10677.06 A10677.14 A10677.22
A10677.06EA 46.3 EUR
A10677.06 A10677.14 A10677.22
Phenylglyoxal monohydrate 97%
Phenylglyoxal monohydrate

Purity: 97% CAS No.: 1075-06-5 Molecular Formula: C8H6O2.H2O Formula Weight: 152.15 (134.12anhy) Melting Point: 71-77° Boiling Point: 142°/125mm Beilstein Reference: 1854721 MDL No.: MFCD00149499

Formel: C₈H₆O₂·H₂O
Siedepunkt: 142 °C (125 mmHg)
Schmelzpunkt: 71…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149499
CAS-Nummer: 1075-06-5
EINECS: 214-036-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR