D(+)-Prolin ≥99%

Lieferant: Thermo Fisher Scientific

A10682.03 A10682.06 A10682.14
A10682.03EA 49.5 EUR
A10682.03 A10682.06 A10682.14
D(+)-Prolin ≥99%
D(+)-Prolin

Purity: 99% CAS No.: 344-25-2 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 213-215° dec. Beilstein Reference: 80811

Formel: C₅H₉NO₂
Dichte: 1,36
Lagertemperatur: Raumtemperatur
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR