DL-Weinsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2R,3R)-rel-2,3-Dihydroxysuccinic acid, Racemic Acid
A10683.0B
A10683.30
A10683.36
A10683.0BEA
159
EUR
A10683.0B
A10683.30
A10683.36
DL-Weinsäure 99%
DL-Tartaric acid
Purity: 99%
CAS No.: 133-37-9
Molecular Formula: C4H6O6
Formula Weight: 150.09
Melting Point: 210-212° dec.
Beilstein Reference: 1725148
MDL No.: MFCD00071626
Formel:
C₄H₆O₆ Molecular Weight: 150.09 g/mol Schmelzpunkt: 210…212 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151254 CAS-Nummer: 133-37-9 EINECS: 205-105-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...