4-Fluor-2-nitroanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10685.18
A10685.22
A10685.36
A10685.18EA
60
EUR
A10685.18
A10685.22
A10685.36
4-Fluor-2-nitroanilin ≥98%
4-Fluor-2-nitroanilin
Purity: 98%
CAS No.: 364-78-3
Molecular Formula: C6H5FN2O2
Formula Weight: 156.12
Flash Point: 89°(192°F)
Melting Point: 90-94°
Beilstein Reference: 2210197
MDL No.: MFCD00007830
Formel:
C₆H₅FN₂O₂ Schmelzpunkt: 90…94 °C Flammpunkt: 89 °C (192 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007830 CAS-Nummer: 364-78-3 EINECS: 206-666-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...