4-Fluor-2-nitroanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10685.18 A10685.22 A10685.36
A10685.18EA 60 EUR
A10685.18 A10685.22 A10685.36
4-Fluor-2-nitroanilin ≥98%
4-Fluor-2-nitroanilin

Purity: 98% CAS No.: 364-78-3 Molecular Formula: C6H5FN2O2 Formula Weight: 156.12 Flash Point: 89°(192°F) Melting Point: 90-94° Beilstein Reference: 2210197 MDL No.: MFCD00007830

Formel: C₆H₅FN₂O₂
Molecular Weight: 156.12 g/mol
Schmelzpunkt: 90…94 °C
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007830
CAS-Nummer: 364-78-3
EINECS: 206-666-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR