Benzil dioxime 98%

Lieferant: Thermo Fisher Scientific

A10687.14 A10687.22
A10687.14EA 61.4 EUR
A10687.14 A10687.22
Benzil dioxime 98%
Benzil dioxime

Purity: 98% CAS No.: 23873-81-6 Molecular Formula: C14H12N2O2 Formula Weight: 240.26 Melting Point: ca 238° dec. Beilstein Reference: 2053615 MDL No.: MFCD00002113

Formel: C₁₄H₁₂N₂O₂
Schmelzpunkt: ∼ 238 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002113
CAS-Nummer: 23873-81-6
EINECS: 245-921-0
Merck Index: 14,1079

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR