Lead(II) citrate trihydrate 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10701.22
A10701.22EA
47.7
EUR
A10701.22
Lead(II) citrate trihydrate 99%
Lead(II) citrate trihydrate
Purity: 99%
CAS No.: 6107-83-1
Molecular Formula: C12H10O14Pb3.3H2O
Formula Weight: 1053.82 (999.77anhy)
Solubility: Soluble in cold water
UN No.: UN2291
MDL No.: MFCD00150456
Formel:
C₁₂H₁₆O₁₇Pb₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150456 CAS-Nummer: 6107-83-1 EINECS: 208-141-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...