ω-Brom-3-methoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10716.06 A10716.14
A10716.06EA 52.3 EUR
A10716.06 A10716.14
ω-Brom-3-methoxyacetophenon ≥98%
ω-Brom-3-methoxyacetophenon

Purity: 98% CAS No.: 5000-65-7 Molecular Formula: C9H9BrO2 Formula Weight: 229.08 Storage Temperature: 2°C to 8°C Odour: Irritating Beilstein Reference: 510128 UN No.: 3261 MDL No.: MFCD00000199

Formel: C₉H₉BrO₂
Siedepunkt: 285 °C (1013 hPa)
Schmelzpunkt: 60…64 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000199
CAS-Nummer: 5000-65-7
EINECS: 225-666-1
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR