3-Hydroxy-4-nitrobenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10724.06 A10724.14 A10724.22
A10724.06EA 21.8 EUR
A10724.06 A10724.14 A10724.22
3-Hydroxy-4-nitrobenzoesäure ≥98%
3-Hydroxy-4-nitrobenzoic acid

Purity: 98% CAS No.: 619-14-7 Molecular Formula: C7H5NO5 Formula Weight: 183.12 Melting Point: 232-236° Beilstein Reference: 2107564

Formel: C₇H₅NO₅
Molecular Weight: 183.12 g/mol
Schmelzpunkt: 232…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007110
CAS-Nummer: 619-14-7
EINECS: 210-580-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR