m-Kresol ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A10733.30
A10733.30EA 31.7 EUR
A10733.30
m-Kresol ≥99%
m-Kresol

Purity: 99% CAS No.: 108-39-4 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 86°(186°F) Refractive Index: 1.5400 Odour: Phenol-like pH: 5 Melting Point: 11-12° Boiling Point: 202-204° Solubility: Soluble in water, alcohol, ether, chloroform Beilstein Reference: 506719 Merck Reference: 14,2579 UN No.: 2076 MDL No.: MFCD00002302

Formel: CH₃C₆H₄OH
Siedepunkt: 202…204 °C
Schmelzpunkt: 11…12 °C
Dichte: 1,034
Flammpunkt: 86 °C (186 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002302
CAS-Nummer: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1,II
Merck Index: 14,2579
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR