Agar, powder
Lieferant: Thermo Fisher Scientific
Synonyme:
Agar Agar
A10752.0E
A10752.22
A10752.36
A10752.0EEA
591
EUR
A10752.0E
A10752.22
A10752.36
Agar, powder
Agar
CAS No.: 9002-18-0
Molecular Formula: (C12H18O9)X
Physical Form: Powder
Appearance: White to cream
Odour: Odourless
Merck Reference: 14,184
MDL No.: MFCD00081288
Schmelzpunkt:
90 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081288 CAS-Nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 14,184 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...