3,4-Pyridinedicarboxylic acid anhydride 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Furo[3,4-c]pyridine-1,3-dione

A10771.14 A10771.03 A10771.06
A10771.14EA 428 EUR
A10771.14 A10771.03 A10771.06
3,4-Pyridinedicarboxylic acid anhydride 97%
3,4-Pyridinedicarboxylic acid anhydride

Purity: 97% CAS No.: 4664-08-8 Molecular Formula: C7H3NO3 Formula Weight: 149.11 Melting Point: 75-79° MDL No.: MFCD00010680

Formel: C₇H₃NO₃
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010680
CAS-Nummer: 4664-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR