Ethyl-2-amino-4-methyl-3-thiophencarboxylat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl 2-amino-4-methylthiophene-3-carboxylate
A10790.06
A10790.14
A10790.06EA
93.8
EUR
A10790.06
A10790.14
Ethyl-2-amino-4-methyl-3-thiophencarboxylat ≥98%
Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Purity: 98+%
CAS No.: 43088-42-2
Molecular Formula: C8H11NO2S
Formula Weight: 185.25
Melting Point: 79-83°
Beilstein Reference: 1366073
Formel:
C₈H₁₁NO₂S Schmelzpunkt: 79…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051669 CAS-Nummer: 43088-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...