Ethyl-2-amino-4-methyl-3-thiophencarboxylat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Ethyl 2-amino-4-methylthiophene-3-carboxylate

A10790.06 A10790.14
A10790.06EA 93.8 EUR
A10790.06 A10790.14
Ethyl-2-amino-4-methyl-3-thiophencarboxylat ≥98%
Ethyl-2-amino-4-methyl-3-thiophencarboxylat

Purity: 98+% CAS No.: 43088-42-2 Molecular Formula: C8H11NO2S Formula Weight: 185.25 Melting Point: 79-83° Beilstein Reference: 1366073

Formel: C₈H₁₁NO₂S
Molecular Weight: 185.25 g/mol
Schmelzpunkt: 79…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051669
CAS-Nummer: 43088-42-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR