4-Fluor-3-nitrobenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10800.06 A10800.14 A10800.22
A10800.06EA 36.1 EUR
A10800.06 A10800.14 A10800.22
4-Fluor-3-nitrobenzoesäure ≥98%
4-Fluoro-3-nitrobenzoic acid

Purity: 98% CAS No.: 453-71-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 121-126° Boiling Point: 92°/15mm MDL No.: MFCD00007058

Formel: C₇H₄FNO₄
Siedepunkt: 92 °C (15 mmHg)
Schmelzpunkt: 121…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007058
CAS-Nummer: 453-71-4
EINECS: 207-221-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR