N-Boc-L-tyrosin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: BOC-L-Tyrosine, Boc-Tyr-OH

A10810.03 A10810.06 A10810.14
A10810.03EA 18 EUR
A10810.03 A10810.06 A10810.14
N-Boc-L-tyrosin ≥98%
N-Boc-L-tyrosin

Purity: 98+% CAS No.: 3978-80-1 Molecular Formula: C14H19NO5 Formula Weight: 281.31 Storage Temperature: 2°C to 8°C Melting Point: 141° dec. Beilstein Reference: 2816406 Optical Rotation: +37° (c=1 in dioxan) MDL No.: MFCD00037179

Formel: C₁₄H₁₉NO₅
Schmelzpunkt: 141 °C (Zersetzung)
MDL: MFCD00037179
CAS-Nummer: 3978-80-1
EINECS: 223-613-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR