(±)-Ethyl-2-brompropionat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate
A10820.18
A10820.30
A10820.36
A10820.18EA
26
EUR
A10820.18
A10820.30
A10820.36
(±)-Ethyl-2-brompropionat ≥98%
(±)-Ethyl-2-brompropionat
Purity: 98+%
CAS No.: 535-11-5
Molecular Formula: C5H9BrO2
Formula Weight: 181.03
Flash Point: 51°(123°F)
Refractive Index: 1.4460
Boiling Point: 158-160°
Beilstein Reference: 773920
Merck Reference: 14,3772
UN No.: 1992
MDL No.: MFCD00000144
Formel:
C₅H₉BrO₂ Molecular Weight: 181.03 g/mol Siedepunkt: 158…160 °C Dichte: 1.391 Flammpunkt: 51 °C (123 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000144 CAS-Nummer: 535-11-5 EINECS: 208-609-5 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...